XBridge BEH Amide XP Column, 130Å, 2.5µm, 2.1mm x 150mm 1 * 1 items
Catalog # WATE186006724
Supplier: WATERS
XBridge BEH Amide XP Column, 130Å, 2.5µm, 2.1mm x 150mm 1 * 1 items
Catalog # WATE186006724
Supplier: WATERS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Crimp caps for crimp neck vials, N 20
Supplier: MACHEREY-NAGEL