Order Entry
Ireland
ContactUsLinkComponent
GLASSWARE WASHER+COND.1PH GW1160SC washing up to 95C - antibacterical thermal disinfection electronic programming: 9 setted programmes + 6 free programmes microprocessor programmable electronic control serial port RS232 for connection to external pr 1 * 1 items
Catalog # SMEGGW1160SC-1
Supplier:  SMEG
GLASSWARE WASHER+COND.1PH GW1160SC washing up to 95C - antibacterical thermal disinfection electronic programming: 9 setted programmes + 6 free programmes microprocessor programmable electronic control serial port RS232 for connection to external pr 1 * 1 items
Catalog # SMEGGW1160SC-1
Supplier:  SMEG
Frequently Bought Together