LDPE bags, clear, compatible for use in cleanroom. Thickness 2mil (50µm), Width 12 (305mm), Length 20 (508mm). Cleanliness level 100 as per NASA JPAG 5322.1, NASA KSC-C-123J, IEST-CC-1246D. 100 pcs per double-pack 1 * 1.000 items
Catalog # FISHPCII12X20X2M_U
Supplier: FISHER CONTAINER
Frequently Bought Together