Order Entry
Ireland
ContactUsLinkComponent
Lid for quartz inceneration crucible per Rademacher in application temperatures of up to 1050C, transparent, high chemical resista nce and purity. Diameter 27mm, height 45mm. The durability or operating cycles of our quartz glass product mainly depe 1 * 1 items
Catalog # TSLA75703
Lid for quartz inceneration crucible per Rademacher in application temperatures of up to 1050C, transparent, high chemical resista nce and purity. Diameter 27mm, height 45mm. The durability or operating cycles of our quartz glass product mainly depe 1 * 1 items
Catalog # TSLA75703
Frequently Bought Together