Order Entry
Ireland
ContactUsLinkComponent
Quartz inceneration crucible per Rademacher in application temperatures of up to 1050C, transparent, high chemical resistance and purity. Diameter 27mm, height 45mm. The durability or operating cycles of our quartz glass product mainly depends on ap 1 * 1 items
Catalog # TSLA75702
Quartz inceneration crucible per Rademacher in application temperatures of up to 1050C, transparent, high chemical resistance and purity. Diameter 27mm, height 45mm. The durability or operating cycles of our quartz glass product mainly depends on ap 1 * 1 items
Catalog # TSLA75702
Frequently Bought Together