Order Entry
Ireland
ContactUsLinkComponent
Trisodium Salt Monohydrate Crystalline Purity: 98% Actually a physical blend of EDTA disodium and EDTA tetrasodium. 1 * 100 g
Catalog # ICNA0215108880
Supplier:  MP Biomedicals
Trisodium Salt Monohydrate Crystalline Purity: 98% Actually a physical blend of EDTA disodium and EDTA tetrasodium. 1 * 100 g
Catalog # ICNA0215108880
Supplier:  MP Biomedicals
Frequently Bought Together