Order Entry
Ireland
ContactUsLinkComponent
ALCOHOL DEHYDROGENASE 2× CRYSTALLINE. From YeastE.C. 1.1.1.1 2X CrystallizedLyophilizedActivity: 300 units/mg protein Unit Definitio n: One unit will convert 1.0 micromole of NAD/min at 25°C 1 * 30 KU
Catalog # ICNA0210016130
Supplier:  MP Biomedicals
ALCOHOL DEHYDROGENASE 2× CRYSTALLINE. From YeastE.C. 1.1.1.1 2X CrystallizedLyophilizedActivity: 300 units/mg protein Unit Definitio n: One unit will convert 1.0 micromole of NAD/min at 25°C 1 * 30 KU
Catalog # ICNA0210016130
Supplier:  MP Biomedicals
Frequently Bought Together