Order Entry
Ireland
ContactUsLinkComponent
SC 150-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC150-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB 1 * 1 items
Catalog # HAKA1533051
SC 150-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC150-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB 1 * 1 items
Catalog # HAKA1533051
Frequently Bought Together