Order Entry
Ireland
ContactUsLinkComponent
SC 100-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC100-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1523051
SC 100-S5P (230V/50Hz) PLASTIC HEATING BATH CIRCULATOR, SC100-S5P, Amb+13 to 100 C, 3-5 L VOLUME, PLASTIC BATH, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
Catalog # HAKA1523051
Frequently Bought Together