Order Entry
Ireland
ContactUsLinkComponent
AC 150-S49 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S49, Amb+13 to 150 C, 28-49 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) & LONG SHAFT REMOTE SENSOR PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1551491
AC 150-S49 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, AC150-S49, Amb+13 to 150 C, 28-49 L BATH VOLUME, 2.0 kW HEATER, FORCE SUCTION PUMP 20 L/MIN & 474 MBAR(330 MBAR SUCTION) & LONG SHAFT REMOTE SENSOR PORT 8 LANGUAGES 1 * 1 items
Catalog # HAKA1551491
Frequently Bought Together