Order Entry
Ireland
ContactUsLinkComponent
pH combination electrode with integral PT1000 T-sensor, gel-filled, pH 2…14, 0 … 60°C open ring diaphragm, Glass body, 1 m cable, MP5 connector 1 * 1 items
Catalog # HACHLZW5053T97002
Supplier:  Hach
pH combination electrode with integral PT1000 T-sensor, gel-filled, pH 2…14, 0 … 60°C open ring diaphragm, Glass body, 1 m cable, MP5 connector 1 * 1 items
Catalog # HACHLZW5053T97002
Supplier:  Hach
Frequently Bought Together