Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

FeTPPS ≥95%

250 mg
  CAYM17187-250
FeTPPS ≥95%
  CAYM17187-250
 :  90384-82-0
FeTPPS ≥95%

250 mg

 :  90384-82-0
 
308,00 €
   
1 * 250 mg
 

FeTPPS (Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride) is a ferric porphyrin complex that causes the decomposition of peroxynitrite by catalytic isomerization to produce nitrate both in vitro and in vivo.

  • Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride

The conversion of this reactive nitrogen species to nitrate results in cytoprotection (EC50 = 5 µM). FeTPPS does not complex with NO and does not alter superoxide directly. It is commonly used to elucidate the roles of peroxynitrite in oxidative stress, cell damage, and intracellular signaling.

Peroxynitrite is a highly reactive nitrogen species formed from the reaction of nitric oxide (NO) and superoxide.

 :  5,10,15,20-Tetrakis(4-sulphonatophenyl)porphyrinato Iron (III), Chloride
  •  :90384-82-0
  •  :C₄₄H₂₈ClFeN₄O₁₂S₄
  •  :MFCD00216780
  •  :1024.27 g/mol
 

 

  • Purity:
    ≥95%
  • Formulation:
    A Crystalline Solid
  • UV/Vis:
    λmax: 204, 324, 396, 530 nm
  • Stability:
    ≥2 years at −20 °C

 

  • Pk:
    250 mg