Order Entry
Ireland
Orders LinkContactUsLinkComponent
FeTPPS ≥95%

FeTPPS ≥95%

100 mg
Catalog # CAYM17187-100
Supplier:   Cayman Chemical
FeTPPS ≥95%
Catalog # CAYM17187-100
Supplier:  Cayman Chemical
CAS Number:  90384-82-0
FeTPPS ≥95%

100 mg

CAS Number:  90384-82-0
Price
160,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

FeTPPS (Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride) is a ferric porphyrin complex that causes the decomposition of peroxynitrite by catalytic isomerization to produce nitrate both in vitro and in vivo.

  • Fe(III)5,10,15,20-tetrakis(4-sulfonatophenyl)porphyrinato chloride

The conversion of this reactive nitrogen species to nitrate results in cytoprotection (EC50 = 5 µM). FeTPPS does not complex with NO and does not alter superoxide directly. It is commonly used to elucidate the roles of peroxynitrite in oxidative stress, cell damage, and intracellular signaling.

Peroxynitrite is a highly reactive nitrogen species formed from the reaction of nitric oxide (NO) and superoxide.

Synonyms:  5,10,15,20-Tetrakis(4-sulphonatophenyl)porphyrinato Iron (III), Chloride
  • CAS Number:90384-82-0
  • Formula:C₄₄H₂₈ClFeN₄O₁₂S₄
  • MDL Number:MFCD00216780
  • Molecular Weight:1024.27 g/mol
Specifications

Specification Test Results

  • Purity:
    ≥95%
  • Formulation:
    A Crystalline Solid
  • UV/Vis:
    λmax: 204, 324, 396, 530 nm
  • Stability:
    ≥2 years at −20 °C

Specifications

  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.