Order Entry
Ireland
ContactUsLinkComponent
An orally bioavailable inhibitor of MEK1/2 (IC50 = 12 nM) that inhibits ERK phosphorylation in various cancer cell lines (IC50s 5 nM) 1 * 50 mg
Catalog # CAYM16996-50
Supplier:  Cayman Chemical
An orally bioavailable inhibitor of MEK1/2 (IC50 = 12 nM) that inhibits ERK phosphorylation in various cancer cell lines (IC50s 5 nM) 1 * 50 mg
Catalog # CAYM16996-50
Supplier:  Cayman Chemical
Frequently Bought Together