Analog I/O interface module VACUUBUS,converts an input signal 4 - 20 mA into a VACUUBUS signal, converts a VACUUBUS signal into a n output signal 0 - 10 V, with 2 m VACUUBUS cable 1 * 1 items
Catalog # VACU20635425
Supplier: VACUUBRAND
Frequently Bought Together