Order Entry
Ireland
ContactUsLinkComponent
SONOREX SUPER RK 514, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 325X300X150 MM 13,5 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAI N BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 860 W, HF-OUTPUT: 215 WEFF. 1 * 1 items
Catalog # BAND277-GB
SONOREX SUPER RK 514, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 325X300X150 MM 13,5 L, S/S TIMER 1-15 MIN AND CONTINUOUS, DRAI N BALL COCK 1/2 , ULTRASONIC PEAK OUTPUT 860 W, HF-OUTPUT: 215 WEFF. 1 * 1 items
Catalog # BAND277-GB
Frequently Bought Together