SONOREX TECHNIK RM 40 UH, ULTRASONIC BATH, 40 KHZ, 230 V 50/60 HZ, ID 480X300X300 MM 45 L, WELDED TANK 2 MM S/S AISI 316 TI TIME R 1-15 MIN AND CONTINUOUS, HEATING 30-80C, ULTRASONIC PEAK OUTPUT 2000 W, HF-OUTPUT: 1X500 WEFF., WITH OVERFLOW GUTT 1 * 1 items
Catalog # BAND8210-CH
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together