ultrasonic bath with power settings, 35 kHz, 230 V( 10%) 50/60 Hz, LED-display for time (1-30 min or continuous). temperature (20 - 80C thermostatically adjustable), power (20 - 100 %). DEGAS-function. ID 325x300x200 mm 18,7 l, s/s ultrasonic 1 * 1 items
Catalog # BAND7185-CH
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together