Order Entry
Ireland
ContactUsLinkComponent
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in weste rn blot. 1 * 1 SET
Catalog # ABNOH00003172-PW2
Supplier:  Abnova
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in weste rn blot. 1 * 1 SET
Catalog # ABNOH00003172-PW2
Supplier:  Abnova
Frequently Bought Together