Order Entry
Ireland
ContactUsLinkComponent
alpha-Mannosidase Assay Kit (Colorimetric) is used for quantitative determination of alpha-Mannosidase activity using colourmetric m ethods. 1 * 1 KIT
Catalog # ABNOKA4566
Supplier:  Abnova
alpha-Mannosidase Assay Kit (Colorimetric) is used for quantitative determination of alpha-Mannosidase activity using colourmetric m ethods. 1 * 1 KIT
Catalog # ABNOKA4566
Supplier:  Abnova
Frequently Bought Together