Order Entry
Ireland
ContactUsLinkComponent
For fast and effective pre-crushing of very hard, hard, medium-hard, abrasive, brittle materials even ferrous alloys. Areas of application Mining and metallurgy, chemistry, geology and mineralogy, glass industry, ceramics, rocks and soils. Method of 1 * 1 items
Catalog # FRIT01.5040.00
Supplier:  FRITSCH
For fast and effective pre-crushing of very hard, hard, medium-hard, abrasive, brittle materials even ferrous alloys. Areas of application Mining and metallurgy, chemistry, geology and mineralogy, glass industry, ceramics, rocks and soils. Method of 1 * 1 items
Catalog # FRIT01.5040.00
Supplier:  FRITSCH
Frequently Bought Together