*NOT FOR INDIVIDUAL SALE USE KIT P/N VHG-XPE502KIT* SCANDIUM - SC @ 1000 µG/G IN HYDROCARBON OIL 1 * 1 items
PROOG-OSC-1000-10G
*NOT FOR INDIVIDUAL SALE USE KIT P/N VHG-XPE502KIT* SCANDIUM - SC @ 1000 µG/G IN HYDROCARBON OIL 1 * 1 items
PROOG-OSC-1000-10G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection