Calcium bromide hydrate for analysis
Plastic bottle, 2,5 kg
Supplier: Thermo Fisher Scientific
Calcium bromide hydrate for analysis
Catalog # ACRO300840025
Supplier: Thermo Fisher Scientific
CAS Number: 71626-99-8
Jump to:
Product Details & Documents
- CAS Number:71626-99-8
- Formula:Br₂Ca·H₂O
- MDL Number:MFCD00149608
- Molecular Weight:217.9 g/mol
- Melting Pt:730 °C
- Density:2,295 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-164-6
Documents
ACRO30084
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder and/or chunks
- Titration Complexometric:≥83.0 % (CaBr2)
- Acidity/alkalinity:Passes test
- Chloride (Cl):≤2000 ppm
- Sulfate (SO4):≤100 ppm
- Bromate and Chlorate:Passes test
- Arsenic (As):≤2 ppm
- Barium (Ba):Passes test
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Subst not pptd:≤0.4 % By ammonium oxalate (as SO4)
Specifications
- Pack type:Plastic bottle
- Pk:2,5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



