MOPS-Na 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, MOPS sodium salt
- CAS Number:71119-22-7
- Formula:C₇H₁₄NNaO₄S
- MDL Number:MFCD00064350
- Molecular Weight:231.25 g/mol
- Storage Temperature:Ambient
- EINECS:428-420-3
Documents
ACRO35259
SDS
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Solubility:(23.125 g/l in cold water) Clear and colorless
- Titration with HClO4:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



