4-Methylindole 99%
Glass bottle, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Methyl-1H-indole
- CAS Number:16096-32-5
- Formula:C₉H₉N
- MDL Number:MFCD00005668
- Molecular Weight:131.18 g/mol
- Boiling Pt:267 °C (760 mmHg)
- Melting Pt:5 °C
- Density:1,06 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Freezer
- EINECS:240-262-5
- UN:3334
- ADR:9,III
Documents
ACRO13389
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid after melting
- Infrared spectrum:Conforms
- Refractive index:1.6060 to 1.6080 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



