Hydroxyurea 98%
Glass bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
- CAS Number:127-07-1
- Formula:CH₄N₂O₂
- MDL Number:MFCD00007943
- Molecular Weight:76.05 g/mol
- Merck Index:13,04874
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:142…146 °C (dec.)
- Storage Temperature:Refrigerator
- EINECS:204-821-7
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:142 to 146 °C
- Titration Iodometric:≥97.5 % (on dried substance)
- Loss on drying:≤1 % (1 g, 60 °C) (< 0.7 kPa)
Specifications
- Pack type:Glass bottle
- Pk:100 g



