COBALT(II)CHLORIDE 1 * 50 mL
Catalog # WAKO031-19041
Supplier: Wako
Frequently Bought Together
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Disodium hydrogen phosphate dodecahydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium carbonate ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium thiocyanate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals