Order Entry
Ireland
ContactUsLinkComponent
environmental cabinet, tall cabinet with wing door(s), model line e-line, model class e-psm, model ep.195.095 dimensions: exterior: wxdxh (mm) 950 x 500 x 1950 interior: wxdxh (mm) 945 x 460 x 1843 body: ral 6016, plain door(s): ral 6016, plain 1 * 1 items
Catalog # MSGA30630009
Supplier:  asecos
environmental cabinet, tall cabinet with wing door(s), model line e-line, model class e-psm, model ep.195.095 dimensions: exterior: wxdxh (mm) 950 x 500 x 1950 interior: wxdxh (mm) 945 x 460 x 1843 body: ral 6016, plain door(s): ral 6016, plain 1 * 1 items
Catalog # MSGA30630009
Supplier:  asecos
Frequently Bought Together