Order Entry
Ireland
ContactUsLinkComponent
grid, as storage level, steel, galvanized load capacity (evenly distributed load) (kg): 180 for medium and tall cabinets with width 1200 mm for model(s): s90, q90 1 * 1 items
Catalog # MSGA27566
Supplier:  asecos
grid, as storage level, steel, galvanized load capacity (evenly distributed load) (kg): 180 for medium and tall cabinets with width 1200 mm for model(s): s90, q90 1 * 1 items
Catalog # MSGA27566
Supplier:  asecos
Frequently Bought Together