Order Entry
Ireland
ContactUsLinkComponent
MABSol® Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax® Human VEGF165, Tag Fre 1 * 25 µG
Catalog # ACRBVE5-H8210-25UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human VEGF165, epitope tag free, primary amine labeling (VE5-H8210) is expressed from human HEK293 cells. It c ontains AA Ala 27 - Arg 191 (Accession NP001165097). It is the biotinylated form of ActiveMax® Human VEGF165, Tag Fre 1 * 25 µG
Catalog # ACRBVE5-H8210-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together