Order Entry
Ireland
ContactUsLinkComponent
PBST is commonly used as a wash solution for Western blot membranes and microtiter plate wells in ELISA assays. The Phosphate Buffer ed Saline with Tween 20 is an optimal formulation of pH stabilizers,salts and detergents designed to effectively remo 1 * 1 L
Catalog # GENOR044
Supplier:  G-Biosciences
PBST is commonly used as a wash solution for Western blot membranes and microtiter plate wells in ELISA assays. The Phosphate Buffer ed Saline with Tween 20 is an optimal formulation of pH stabilizers,salts and detergents designed to effectively remo 1 * 1 L
Catalog # GENOR044
Supplier:  G-Biosciences
Frequently Bought Together