Order Entry
Ireland
ContactUsLinkComponent
G-Biosciences scientists have developed a high efficiency transfer buffer for Western blotting. The pre-made and ready-to-use [20X] transfer buffer is prepared for optimal conductivity and efficient protein transfer without generating excessive heat 1 * 1 gal US
Catalog # GENO786-019G
Supplier:  G-Biosciences
G-Biosciences scientists have developed a high efficiency transfer buffer for Western blotting. The pre-made and ready-to-use [20X] transfer buffer is prepared for optimal conductivity and efficient protein transfer without generating excessive heat 1 * 1 gal US
Catalog # GENO786-019G
Supplier:  G-Biosciences