Order Entry
Ireland
ContactUsLinkComponent
Citraconic anhydride reacts with primary amines and blocks them by creating an amide linkage and a terminal carboxylate.The linkage is stable at neutral to alkaline pH (pH >7) and at acidic conditions (pH 4) the amide linkage is rapidly hydrolyzed to 1 * 1 KIT
Catalog # GENO786-389
Supplier:  G-Biosciences
Citraconic anhydride reacts with primary amines and blocks them by creating an amide linkage and a terminal carboxylate.The linkage is stable at neutral to alkaline pH (pH >7) and at acidic conditions (pH 4) the amide linkage is rapidly hydrolyzed to 1 * 1 KIT
Catalog # GENO786-389
Supplier:  G-Biosciences