Order Entry
Ireland
ContactUsLinkComponent
Plcd3 - Mouse, 4 unique 29mer shRNA constructs in retroviral RFP vector (Gene ID = 72469). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pRFP-C-RS Vector, TR30015, included for free. 1 * 1 KIT
Catalog # ORIGTF504740
Supplier:  OriGene
Plcd3 - Mouse, 4 unique 29mer shRNA constructs in retroviral RFP vector (Gene ID = 72469). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pRFP-C-RS Vector, TR30015, included for free. 1 * 1 KIT
Catalog # ORIGTF504740
Supplier:  OriGene
Frequently Bought Together