Methylene blue solution
3×1 l, 3
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
- CAS Number:61-73-4
- Formula:C₁₆H₁₈ClN₃S
- MDL Number:MFCD00150008
- Molecular Weight:319.86 g/mol
Documents
Certificates
Search by Lot Number911490ZA_METHYLENE BLUE SOLUTION_ISO 11133_rev1.pdf 911490ZAMP_METHYLENE BLUE SOLUTION_ISO 11133_rev1.pdf
spec_VWRC911490ZA
Product specification
VWR Microbiology
Product Literature
Specifications
Specifications
- Pk:3
- Pk Info:3×1 l
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



