Quartz glass muffle tray in application temperatures of up to 1050C (maximum 1600C), 4 holes, rod type, translucent, high chemica l resistance and purity. Length 76mm, height 76mm. 1 * 1 items
Catalog # TSLA73171
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together