Order Entry
Ireland
ContactUsLinkComponent
E3 INCUBATOR ,,50 litres capacity ,,Temperature range ambient +5 to 70°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 40 high x 36 wide x 35 deep ,,External dimensions (cms): 66 1 * 1 items
  GENLE3/INC/50
 :  GENLAB
E3 INCUBATOR ,,50 litres capacity ,,Temperature range ambient +5 to 70°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 40 high x 36 wide x 35 deep ,,External dimensions (cms): 66 1 * 1 items
  GENLE3/INC/50
 :  GENLAB
Frequently Bought Together