Order Entry
Ireland
ContactUsLinkComponent
E3 DRYING OVEN ,,30 litres capacity ,,Temperature range 40 to 250°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 24 high x 36 wide x 35 deep ,,External dimensions (cms): 50 high 1 * 1 items
Catalog # GENLE3/OVN/30
Supplier:  GENLAB
E3 DRYING OVEN ,,30 litres capacity ,,Temperature range 40 to 250°C ,,Fluctuation ± 0.25°C ,,Stainless steel interior ,,Easy clean powder coated body ,,Internal dimensions (cms): 24 high x 36 wide x 35 deep ,,External dimensions (cms): 50 high 1 * 1 items
Catalog # GENLE3/OVN/30
Supplier:  GENLAB
Frequently Bought Together