Order Entry
Ireland
ContactUsLinkComponent
Bottles glass ointment jar white flint 250ml type III soda lime glass with fitted R3/83 natural PP wadless caps max height 73.80mm max dia 87.00mm 1 * 10 items
Catalog # 215-3358
Supplier:  Azpack
undefined
Bottles glass ointment jar white flint 250ml type III soda lime glass with fitted R3/83 natural PP wadless caps max height 73.80mm max dia 87.00mm 1 * 10 items
Catalog # 215-3358
Supplier:  Azpack
Frequently Bought Together