Order Entry
Ireland
ContactUsLinkComponent
POLARIMETER ADP 660 DIGITAL (DEN PLUG) MULTIPLE WAVELENGTH 325,365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0790
Supplier:  BELLINGHAM STANLEY
undefined
POLARIMETER ADP 660 DIGITAL (DEN PLUG) MULTIPLE WAVELENGTH 325,365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0790
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together