Order Entry
Ireland
ContactUsLinkComponent
POLARIMETER ADP 640 DIGITAL (EU PLUG) MULTIPLE WAVELENGTH 405,436,546 & 589NM 1 * 1 items
Catalog # 635-0784
Supplier:  BELLINGHAM STANLEY
undefined
POLARIMETER ADP 640 DIGITAL (EU PLUG) MULTIPLE WAVELENGTH 405,436,546 & 589NM 1 * 1 items
Catalog # 635-0784
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together