Order Entry
Ireland
ContactUsLinkComponent
POLARIMETER ADP 620 DIGITAL (EU PLUG) DUAL WAVELENGTH 546 & 589NM 1 * 1 items
Catalog # 635-0776
Supplier:  BELLINGHAM STANLEY
undefined
POLARIMETER ADP 620 DIGITAL (EU PLUG) DUAL WAVELENGTH 546 & 589NM 1 * 1 items
Catalog # 635-0776
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together