Order Entry
Ireland
ContactUsLinkComponent
BCECF-Acetoxymethyl (BCECF-AM) ester is a widely-used fluorescent indicator for estimating intracellular pH.{25713,25712} This membr ane-permeant compound is modified by intracellular esterases to produce BCECF, a polar fluorescein derivative that is 1 * 1 mg
Catalog # CAYM15922-1
Supplier:  Cayman Chemical
BCECF-Acetoxymethyl (BCECF-AM) ester is a widely-used fluorescent indicator for estimating intracellular pH.{25713,25712} This membr ane-permeant compound is modified by intracellular esterases to produce BCECF, a polar fluorescein derivative that is 1 * 1 mg
Catalog # CAYM15922-1
Supplier:  Cayman Chemical
Frequently Bought Together