Order Entry
Ireland
ContactUsLinkComponent
qPCR detection of Proteus mirabilis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-P.MIRABIL
Supplier:  PRIMERDESIGN
qPCR detection of Proteus mirabilis. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endo genous control, extraction control 1 * 150 Tests
Catalog # PATH-P.MIRABIL
Supplier:  PRIMERDESIGN
Frequently Bought Together