Order Entry
Ireland
ContactUsLinkComponent
qPCR detection of Human Influenza type A M2. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cur ve, Endogenous control, extraction control 1 * 150 Tests
Catalog # ZPATHFLUAM2
Supplier:  PRIMERDESIGN
qPCR detection of Human Influenza type A M2. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard cur ve, Endogenous control, extraction control 1 * 150 Tests
Catalog # ZPATHFLUAM2
Supplier:  PRIMERDESIGN
Frequently Bought Together