Order Entry
Ireland
ContactUsLinkComponent
qPCR detection of Clavibacter michiganensis sub species michiganensis. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-CMM
Supplier:  PRIMERDESIGN
qPCR detection of Clavibacter michiganensis sub species michiganensis. Contains: Primer and probe, RT target specific primers (RNA v iruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-CMM
Supplier:  PRIMERDESIGN
Frequently Bought Together