Order Entry
Ireland
ContactUsLinkComponent
qPCR detection of all Bacteroides species. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve , Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-BACTEROID
Supplier:  PRIMERDESIGN
qPCR detection of all Bacteroides species. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve , Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-BACTEROID
Supplier:  PRIMERDESIGN
Frequently Bought Together