Order Entry
Ireland
ContactUsLinkComponent
Nikon SMZ745T stereo microscope with Hybrid LED stand. Includes: NIKOMMA36410 SMZ745T Zooming Body 2x NIKOMMK30102 C-W 10xB Eyepiece with dioptre adjustment NIKOMMD23500 C-LEDS Hybrid LED Stand NIKO17023471 UK Power Cord 5-amp 1.95m 1 * 1 items
Catalog # 630-2505
Supplier:  NIKON
Nikon SMZ745T stereo microscope with Hybrid LED stand. Includes: NIKOMMA36410 SMZ745T Zooming Body 2x NIKOMMK30102 C-W 10xB Eyepiece with dioptre adjustment NIKOMMD23500 C-LEDS Hybrid LED Stand NIKO17023471 UK Power Cord 5-amp 1.95m 1 * 1 items
Catalog # 630-2505
Supplier:  NIKON
Frequently Bought Together