Order Entry
Ireland
ContactUsLinkComponent
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 50kD 34mm flat width (22mm dia meter) 3,7ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132130T
Dialysis tubing Trial kit Spectra/Por 7 pre-treated standard regenerated cellulose (RC) membrane MWCO 50kD 34mm flat width (22mm dia meter) 3,7ml/cm including tubing closures and openers wet 1 * 1 m
Catalog # SPEC132130T
Frequently Bought Together