Order Entry
Ireland
ContactUsLinkComponent
Millikan’s apparatus, 6mm Plate diameter, 80mm Halogen lamp, 12V/10W Objective magnification, 2x Eyepiece magnification, 10x Microme ter range: 10mm, Scale divisions: 0.1mm. 1 * 1 items
Catalog # BINH1013769
Supplier:  3B Scientific
Millikan’s apparatus, 6mm Plate diameter, 80mm Halogen lamp, 12V/10W Objective magnification, 2x Eyepiece magnification, 10x Microme ter range: 10mm, Scale divisions: 0.1mm. 1 * 1 items
Catalog # BINH1013769
Supplier:  3B Scientific
Frequently Bought Together